Assay Details
| Gene Symbol: Mrpl46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852736.3 | XP_004852793.1 | 1 - 2 | 267 | 88 | |
| XM_021264797.1 | XP_021120456.1 | 1 - 2 | 268 | 88 | ||
More Information
Set Membership: |
|

Mrpl46
Genomic Map