Assay Details
| Gene Symbol: Ggh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256083.1 | XP_021111742.1 | 7 - 8 | 819 | 75 | |
| XM_021256084.1 | XP_021111743.1 | 8 - 9 | 932 | 75 | ||
More Information
Set Membership: |
|

Ggh
Genomic Map