Assay Details
| Gene Symbol: LOC101704668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867034.3 | XP_004867091.1 | 3 - 4 | 579 | 76 | |
| XM_004867035.1 | XP_004867092.1 | 3 - 4 | 579 | 76 | ||
More Information
Set Membership: |
|

LOC101704668
Genomic Map