Assay Details
| Gene Symbol: Tbata | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259563.1 | XP_021115222.1 | 4 - 5 | 286 | 67 | |
| XM_021259564.1 | XP_021115223.1 | 5 - 6 | 337 | 67 | ||
| XM_021259565.1 | XP_021115224.1 | 5 - 6 | 293 | 67 | ||
| XM_021259566.1 | XP_021115225.1 | 4 - 5 | 240 | 67 | ||
| XM_021259567.1 | XP_021115226.1 | 4 - 5 | 286 | 67 | ||
| XM_021259568.1 | XP_021115227.1 | 3 - 4 | 402 | 67 | ||
| XM_021259569.1 | XP_021115228.1 | 4 - 5 | 453 | 67 | ||
| XM_021259570.1 | XP_021115229.1 | 3 - 4 | 356 | 67 | ||
| XM_021259571.1 | XP_021115230.1 | 3 - 4 | 402 | 67 | ||
| XM_021259572.1 | XP_021115231.1 | 4 - 5 | 286 | 67 | ||
More Information
Set Membership: |
|

Tbata
Genomic Map