Assay Details
| Gene Symbol: Acp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868297.3 | XP_004868354.1 | 5 - 6 | 493 | 77 | |
| XM_004868298.3 | XP_004868355.1 | 5 - 6 | 493 | 77 | ||
| XM_021247267.1 | XP_021102926.1 | 6 - 7 | 571 | 77 | ||
| XM_021247268.1 | XP_021102927.1 | 6 - 7 | 571 | 77 | ||
More Information
Set Membership: |
|

Acp1
Genomic Map