Assay Details
| Gene Symbol: Wrap73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863766.2 | XP_004863823.1 | 5 - 6 | 636 | 61 | |
| XM_004863767.2 | XP_004863824.1 | 4 - 5 | 530 | 61 | ||
| XM_021243398.1 | XP_021099057.1 | 5 - 6 | 636 | 61 | ||
More Information
Set Membership: |
|

Wrap73
Genomic Map