Assay Details
| Gene Symbol: LOC101710289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862676.2 | XP_004862733.1 | 14 - 15 | 1549 | 120 | |
| XM_021242757.1 | XP_021098416.1 | 15 - 16 | 1804 | 120 | ||
| XM_021242759.1 | XP_021098418.1 | 14 - 15 | 1680 | 120 | ||
| XM_021242760.1 | XP_021098419.1 | 15 - 16 | 1818 | 120 | ||
More Information
Set Membership: |
|

LOC101710289
Genomic Map