Assay Details
| Gene Symbol: LOC101710289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862676.2 | XP_004862733.1 | 17 - 18 | 1820 | 73 | |
| XM_021242757.1 | XP_021098416.1 | 18 - 19 | 2075 | 73 | ||
| XM_021242758.1 | XP_021098417.1 | 17 - 18 | 2011 | 73 | ||
| XM_021242759.1 | XP_021098418.1 | 17 - 18 | 1951 | 73 | ||
More Information
Set Membership: |
|

LOC101710289
Genomic Map