Assay Details
| Gene Symbol: LOC101710289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862676.2 | XP_004862733.1 | 1 - 2 | 198 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101710289| Gene Symbol: LOC101710289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862676.2 | XP_004862733.1 | 1 - 2 | 198 | 66 | |
Set Membership: |
|