Assay Details
| Gene Symbol: Ercc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862894.3 | XP_004862951.1 | 7 - 8 | 748 | 95 | |
| XM_004862895.3 | XP_004862952.1 | 7 - 8 | 736 | 95 | ||
| XM_004862897.3 | XP_004862954.1 | 7 - 8 | 748 | 95 | ||
| XM_021242979.1 | XP_021098638.1 | 7 - 8 | 748 | 95 | ||
| XM_021242980.1 | XP_021098639.1 | 7 - 8 | 736 | 95 | ||
| XM_021242981.1 | XP_021098640.1 | 7 - 8 | 676 | 95 | ||
| XM_021242982.1 | XP_021098641.1 | 6 - 7 | 611 | 95 | ||
| XM_021242983.1 | XP_021098642.1 | 8 - 9 | 739 | 95 | ||
| XM_021242984.1 | XP_021098643.1 | 8 - 9 | 730 | 95 | ||
| XM_021242985.1 | XP_021098644.1 | 7 - 8 | 748 | 95 | ||
| XM_021242986.1 | XP_021098645.1 | 5 - 6 | 509 | 95 | ||
More Information
Set Membership: |
|

Ercc8
Genomic Map