Assay Details
| Gene Symbol: LOC101709814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863844.2 | XP_004863901.1 | 6 - 7 | 795 | 64 | |
| XM_021243363.1 | XP_021099022.1 | 6 - 7 | 499 | 64 | ||
More Information
Set Membership: |
|

LOC101709814
Genomic Map