Assay Details
| Gene Symbol: Nup35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868182.2 | XP_004868239.1 | 3 - 4 | 500 | 70 | |
| XM_021247116.1 | XP_021102775.1 | 3 - 4 | 463 | 70 | ||
| XM_021247117.1 | XP_021102776.1 | 3 - 4 | 321 | 70 | ||
More Information
Set Membership: |
|

Nup35
Genomic Map