Assay Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861819.3 | XP_004861876.1 | 7 - 8 | 716 | 56 | |
| XM_004861820.3 | XP_004861877.1 | 6 - 7 | 1238 | 56 | ||
| XM_004861821.3 | XP_004861878.1 | 7 - 8 | 1302 | 56 | ||
| XM_021242066.1 | XP_021097725.1 | 7 - 8 | 1309 | 56 | ||
| XM_021242067.1 | XP_021097726.1 | 6 - 7 | 650 | 56 | ||
More Information
Set Membership: |
|

Sec61a2
Genomic Map