Assay Details
| Gene Symbol: Vapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840939.3 | XP_004840996.1 | 1 - 2 | 208 | 81 | |
| XM_004840940.2 | XP_004840997.1 | 1 - 2 | 208 | 81 | ||
More Information
Set Membership: |
|

Vapb
Genomic Map