Assay Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858669.3 | XP_004858726.1 | 7 - 8 | 1110 | 56 | |
| XM_004858670.3 | XP_004858727.1 | 7 - 8 | 1110 | 56 | ||
| XM_004858671.3 | XP_004858728.1 | 5 - 6 | 739 | 56 | ||
| XM_021239606.1 | XP_021095265.1 | 4 - 5 | 690 | 56 | ||
| XM_021239607.1 | XP_021095266.1 | 3 - 4 | 873 | 56 | ||
More Information
Set Membership: |
|

Ppox
Genomic Map