Assay Details
| Gene Symbol: Chst10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844438.2 | XP_004844495.1 | 2 - 3 | 243 | 77 | |
| XM_021258082.1 | XP_021113741.1 | 3 - 4 | 395 | 77 | ||
More Information
Set Membership: |
|

Chst10
Genomic Map