Assay Details
| Gene Symbol: Plpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869632.3 | XP_004869689.1 | 3 - 4 | 1066 | 57 | |
| XM_004869633.3 | XP_004869690.1 | 3 - 4 | 1075 | 57 | ||
More Information
Set Membership: |
|

Plpp3
Genomic Map