Assay Details
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837394.3 | XP_004837451.1 | 7 - 8 | 698 | 60 | |
XM_004837395.3 | XP_004837452.1 | 5 - 6 | 533 | 60 | ||
XM_004837397.3 | XP_004837454.1 | 5 - 6 | 639 | 60 | ||
XM_004837398.3 | XP_004837455.1 | 6 - 7 | 758 | 60 | ||
XM_004837399.3 | XP_004837456.1 | 5 - 6 | 420 | 60 | ||
XM_021247649.1 | XP_021103308.1 | 6 - 7 | 614 | 60 | ||
XM_021247651.1 | XP_021103310.1 | 6 - 7 | 663 | 60 | ||
XM_021247659.1 | XP_021103318.1 | 6 - 7 | 554 | 60 | ||
XM_021247667.1 | XP_021103326.1 | 6 - 7 | 652 | 60 | ||
XM_021247669.1 | XP_021103328.1 | 5 - 6 | 410 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
