Assay Details
| Gene Symbol: LOC101698847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867877.3 | XP_004867934.1 | 6 - 7 | 1868 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101698847| Gene Symbol: LOC101698847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867877.3 | XP_004867934.1 | 6 - 7 | 1868 | 78 | |
Set Membership: |
|