Assay Details
| Gene Symbol: Zmpste24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851048.3 | XP_004851105.1 | 2 - 3 | 467 | 69 | |
| XM_004851049.3 | XP_004851106.1 | 2 - 3 | 469 | 69 | ||
| XM_021263406.1 | XP_021119065.1 | 2 - 3 | 471 | 69 | ||
| XM_021263407.1 | XP_021119066.1 | 2 - 3 | 473 | 69 | ||
| XM_021263408.1 | XP_021119067.1 | 2 - 3 | 274 | 69 | ||
| XM_021263409.1 | XP_021119068.1 | 1 - 2 | 139 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021263409.1 transcripts.
Set Membership: |
|

Zmpste24
Genomic Map