Assay Details
| Gene Symbol: Tp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834790.2 | XP_004834847.1 | 5 - 6 | 901 | 83 | |
| XM_004834791.2 | XP_004834848.1 | 5 - 6 | 901 | 83 | ||
| XM_004834792.2 | XP_004834849.1 | 5 - 6 | 901 | 83 | ||
| XM_021255678.1 | XP_021111337.1 | 5 - 6 | 901 | 83 | ||
| XM_021255679.1 | XP_021111338.1 | 3 - 4 | 585 | 83 | ||
More Information
Set Membership: |
|

Tp63
Genomic Map