Assay Details
| Gene Symbol: Tmem184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845615.2 | XP_004845672.1 | 5 - 6 | 803 | 60 | |
| XM_004845616.2 | XP_004845673.1 | 5 - 6 | 800 | 60 | ||
| XM_004845617.2 | XP_004845674.1 | 4 - 5 | 623 | 60 | ||
| XM_013072570.2 | XP_012928024.1 | 4 - 5 | 1101 | 60 | ||
| XM_021258997.1 | XP_021114656.1 | 5 - 6 | 808 | 60 | ||
| XM_021258998.1 | XP_021114657.1 | 5 - 6 | 805 | 60 | ||
More Information
Set Membership: |
|

Tmem184b
Genomic Map