Assay Details
| Gene Symbol: Riox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077081.2 | XP_012932535.1 | 2 - 3 | 376 | 109 | |
| XM_021238839.1 | XP_021094498.1 | 2 - 3 | 356 | 109 | ||
| XM_021238840.1 | XP_021094499.1 | 3 - 4 | 261 | 109 | ||
| XM_021238841.1 | XP_021094500.1 | 2 - 3 | 222 | 109 | ||
| XM_021238844.1 | XP_021094503.1 | 2 - 3 | 376 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013077081.2, XM_021238839.1, XM_021238840.1, XM_021238841.1, XM_021238844.1 transcripts.
Set Membership: |
|

Riox2
Genomic Map