Assay Details
| Gene Symbol: Riox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857929.3 | XP_004857986.1 | 6 - 7 | 1206 | 80 | |
| XM_004857931.3 | XP_004857988.1 | 6 - 7 | 1226 | 80 | ||
| XM_004857932.3 | XP_004857989.1 | 7 - 8 | 1111 | 80 | ||
| XM_004857933.3 | XP_004857990.1 | 6 - 7 | 1072 | 80 | ||
| XM_013077081.2 | XP_012932535.1 | 5 - 6 | 1101 | 80 | ||
| XM_013077082.1 | XP_012932536.1 | 6 - 7 | 1226 | 80 | ||
| XM_013077083.1 | XP_012932537.1 | 6 - 7 | 1226 | 80 | ||
| XM_013077085.2 | XP_012932539.1 | 7 - 8 | 1044 | 80 | ||
| XM_021238839.1 | XP_021094498.1 | 5 - 6 | 1081 | 80 | ||
| XM_021238840.1 | XP_021094499.1 | 6 - 7 | 986 | 80 | ||
| XM_021238841.1 | XP_021094500.1 | 5 - 6 | 947 | 80 | ||
| XM_021238843.1 | XP_021094502.1 | 6 - 7 | 1226 | 80 | ||
| XM_021238844.1 | XP_021094503.1 | 6 - 7 | 919 | 80 | ||
More Information
Set Membership: |
|

Riox2
Genomic Map