Assay Details
| Gene Symbol: LOC101697672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857237.3 | XP_004857294.1 | 3 - 4 | 669 | 64 | |
| XM_004857238.3 | XP_004857295.1 | 2 - 3 | 531 | 64 | ||
More Information
Set Membership: |
|

LOC101697672
Genomic Map