Assay Details
| Gene Symbol: Mst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834046.2 | XP_004834103.1 | 14 - 15 | 1637 | 59 | |
| XM_021262717.1 | XP_021118376.1 | 12 - 13 | 1349 | 59 | ||
More Information
Set Membership: |
|

Mst1
Genomic Map