Assay Details
Gene Symbol: LOC101713959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068377.2 | XP_012923831.1 | 6 - 7 | 1031 | 85 | |
XM_021250407.1 | XP_021106066.1 | 6 - 7 | 1049 | 85 | ||
XM_021250408.1 | XP_021106067.1 | 7 - 8 | 1085 | 85 | ||
XM_021250409.1 | XP_021106068.1 | 7 - 8 | 1103 | 85 | ||
XM_021250410.1 | XP_021106069.1 | 7 - 8 | 1097 | 85 | ||
XM_021250411.1 | XP_021106070.1 | 6 - 7 | 1019 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
