Assay Details
| Gene Symbol: LOC101713959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068377.2 | XP_012923831.1 | 8 - 9 | 1222 | 93 | |
| XM_021250407.1 | XP_021106066.1 | 8 - 9 | 1240 | 93 | ||
| XM_021250408.1 | XP_021106067.1 | 9 - 10 | 1276 | 93 | ||
| XM_021250409.1 | XP_021106068.1 | 9 - 10 | 1294 | 93 | ||
| XM_021250410.1 | XP_021106069.1 | 9 - 10 | 1288 | 93 | ||
| XM_021250411.1 | XP_021106070.1 | 8 - 9 | 1210 | 93 | ||
More Information
Set Membership: |
|

LOC101713959
Genomic Map