Assay Details
Gene Symbol: LOC101713959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021250407.1 | XP_021106066.1 | 9 - 10 | 1314 | 104 | |
XM_021250408.1 | XP_021106067.1 | 10 - 11 | 1350 | 104 | ||
XM_021250409.1 | XP_021106068.1 | 10 - 11 | 1368 | 104 | ||
XM_021250410.1 | XP_021106069.1 | 10 - 11 | 1362 | 104 | ||
XM_021250411.1 | XP_021106070.1 | 9 - 10 | 1284 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
