Assay Details
| Gene Symbol: Cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076401.2 | XP_012931855.1 | 4 - 5 | 530 | 75 | |
| XM_021237458.1 | XP_021093117.1 | 3 - 4 | 826 | 75 | ||
| XM_021237459.1 | XP_021093118.1 | 4 - 5 | 661 | 75 | ||
| XM_021237460.1 | XP_021093119.1 | 4 - 5 | 413 | 75 | ||
| XM_021237461.1 | XP_021093120.1 | 3 - 4 | 335 | 75 | ||
More Information
Set Membership: |
|

Cd2bp2
Genomic Map