Assay Details
| Gene Symbol: LOC101726271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870750.3 | XP_004870807.1 | 3 - 4 | 636 | 65 | |
| XM_021248989.1 | XP_021104648.1 | 3 - 4 | 636 | 65 | ||
More Information
Set Membership: |
|

LOC101726271
Genomic Map