Assay Details
| Gene Symbol: LOC101726271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870750.3 | XP_004870807.1 | 6 - 7 | 1069 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101726271| Gene Symbol: LOC101726271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870750.3 | XP_004870807.1 | 6 - 7 | 1069 | 92 | |
Set Membership: |
|