Assay Details
| Gene Symbol: Retreg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864555.3 | XP_004864612.1 | 7 - 8 | 973 | 75 | |
| XM_021244260.1 | XP_021099919.1 | 6 - 7 | 505 | 75 | ||
More Information
Set Membership: |
|

Retreg2
Genomic Map