Assay Details
| Gene Symbol: Ripor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260545.1 | XP_021116204.1 | 19 - 20 | 2868 | 75 | |
| XM_021260546.1 | XP_021116205.1 | 19 - 20 | 3267 | 75 | ||
| XM_021260547.1 | XP_021116206.1 | 19 - 20 | 2847 | 75 | ||
| XM_021260548.1 | XP_021116207.1 | 19 - 20 | 2834 | 75 | ||
| XM_021260549.1 | XP_021116208.1 | 19 - 20 | 2785 | 75 | ||
| XM_021260550.1 | XP_021116209.1 | 18 - 19 | 2713 | 75 | ||
| XM_021260551.1 | XP_021116210.1 | 19 - 20 | 2862 | 75 | ||
| XM_021260552.1 | XP_021116211.1 | 18 - 19 | 2708 | 75 | ||
More Information
Set Membership: |
|

Ripor2
Genomic Map