Assay Details
| Gene Symbol: Myl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851114.2 | XP_004851171.1 | 4 - 5 | 584 | 63 | |
| XM_013074461.2 | XP_012929915.1 | 4 - 5 | 453 | 63 | ||
More Information
Set Membership: |
|

Myl1
Genomic Map