Assay Details
| Gene Symbol: Hscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843365.2 | XP_004843422.1 | 1 - 2 | 328 | 75 | |
| XM_021257155.1 | XP_021112814.1 | 1 - 2 | 332 | 75 | ||
| XM_021257156.1 | XP_021112815.1 | 1 - 2 | 333 | 75 | ||
More Information
Set Membership: |
|

Hscb
Genomic Map