Assay Details
| Gene Symbol: Bdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866257.3 | XP_004866314.1 | 5 - 6 | 478 | 85 | |
| XM_021245404.1 | XP_021101063.1 | 6 - 7 | 1502 | 85 | ||
| XM_021245405.1 | XP_021101064.1 | 7 - 8 | 1538 | 85 | ||
| XM_021245406.1 | XP_021101065.1 | 5 - 6 | 1391 | 85 | ||
| XM_021245407.1 | XP_021101066.1 | 6 - 7 | 1501 | 85 | ||
| XM_021245408.1 | XP_021101067.1 | 5 - 6 | 479 | 85 | ||
More Information
Set Membership: |
|

Bdh2
Genomic Map