Assay Details
| Gene Symbol: Galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838443.2 | XP_004838500.1 | 3 - 4 | 422 | 63 | |
| XM_013069673.2 | XP_012925127.1 | 3 - 4 | 378 | 63 | ||
| XM_013069674.2 | XP_012925128.1 | 3 - 4 | 495 | 63 | ||
| XM_021253001.1 | XP_021108660.1 | 2 - 3 | 98 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021253001.1 transcripts.
Set Membership: |
|

Galt
Genomic Map