Assay Details
| Gene Symbol: Mbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861930.3 | XP_004861987.1 | 8 - 9 | 991 | 57 | |
| XM_004861931.3 | XP_004861988.2 | 8 - 9 | 1047 | 57 | ||
| XM_013078552.2 | XP_012934006.1 | 9 - 10 | 1074 | 57 | ||
| XM_013078553.2 | XP_012934007.1 | 7 - 8 | 960 | 57 | ||
More Information
Set Membership: |
|

Mbp
Genomic Map