Assay Details
| Gene Symbol: Pank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021253029.1 | XP_021108688.1 | 5 - 6 | 1820 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pank1| Gene Symbol: Pank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021253029.1 | XP_021108688.1 | 5 - 6 | 1820 | 68 | |
Set Membership: |
|