Assay Details
| Gene Symbol: LOC101705012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846867.2 | XP_004846924.1 | 1 - 2 | 181 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101705012| Gene Symbol: LOC101705012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846867.2 | XP_004846924.1 | 1 - 2 | 181 | 82 | |
Set Membership: |
|