Assay Details
| Gene Symbol: Slc35a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860235.3 | XP_004860292.1 | 3 - 4 | 532 | 75 | |
| XM_021240784.1 | XP_021096443.1 | 3 - 4 | 606 | 75 | ||
More Information
Set Membership: |
|

Slc35a1
Genomic Map