Assay Details
| Gene Symbol: Ldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865556.3 | XP_004865613.1 | 8 - 9 | 1259 | 59 | |
| XM_021244873.1 | XP_021100532.1 | 8 - 9 | 1605 | 59 | ||
| XM_021244874.1 | XP_021100533.1 | 8 - 9 | 1605 | 59 | ||
More Information
Set Membership: |
|

Ldlr
Genomic Map