Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856996.3 | XP_004857053.1 | 18 - 19 | 3347 | 61 | |
| XM_013076730.2 | XP_012932184.1 | 18 - 19 | 3363 | 61 | ||
| XM_021238269.1 | XP_021093928.1 | 18 - 19 | 3333 | 61 | ||
| XM_021238270.1 | XP_021093929.1 | 18 - 19 | 3317 | 61 | ||
More Information
Set Membership: |
|

Spag5
Genomic Map