Assay Details
| Gene Symbol: Lims2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835966.3 | XP_004836023.2 | 8 - 9 | 1146 | 57 | |
| XM_004835967.2 | XP_004836024.1 | 8 - 9 | 893 | 57 | ||
| XM_004835968.2 | XP_004836025.1 | 9 - 10 | 1026 | 57 | ||
| XM_004835969.3 | XP_004836026.1 | 7 - 8 | 703 | 57 | ||
| XM_021266338.1 | XP_021121997.1 | 8 - 9 | 813 | 57 | ||
More Information
Set Membership: |
|

Lims2
Genomic Map