Assay Details
| Gene Symbol: Lin7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851578.2 | XP_004851635.1 | 3 - 4 | 329 | 70 | |
| XM_021263950.1 | XP_021119609.1 | 4 - 5 | 354 | 70 | ||
| XM_021263951.1 | XP_021119610.1 | 4 - 5 | 392 | 70 | ||
| XM_021263952.1 | XP_021119611.1 | 4 - 5 | 379 | 70 | ||
| XM_021263953.1 | XP_021119612.1 | 4 - 5 | 384 | 70 | ||
More Information
Set Membership: |
|

Lin7c
Genomic Map