Assay Details
| Gene Symbol: Naxd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239358.1 | XP_021095017.1 | 4 - 5 | 552 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naxd| Gene Symbol: Naxd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239358.1 | XP_021095017.1 | 4 - 5 | 552 | 52 | |
Set Membership: |
|