Assay Details
| Gene Symbol: Spag6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859872.3 | XP_004859929.1 | 4 - 5 | 942 | 61 | |
| XM_004859873.3 | XP_004859930.1 | 4 - 5 | 931 | 61 | ||
| XM_004859875.3 | XP_004859932.1 | 3 - 4 | 846 | 61 | ||
| XM_004859876.2 | XP_004859933.1 | 4 - 5 | 934 | 61 | ||
| XM_004859878.3 | XP_004859935.1 | 4 - 5 | 949 | 61 | ||
| XM_021240467.1 | XP_021096126.1 | 4 - 5 | 606 | 61 | ||
| XM_021240468.1 | XP_021096127.1 | 3 - 4 | 515 | 61 | ||
| XM_021240469.1 | XP_021096128.1 | 4 - 5 | 941 | 61 | ||
More Information
Set Membership: |
|

Spag6
Genomic Map