Assay Details
| Gene Symbol: Ccnh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841654.2 | XP_004841711.1 | 1 - 2 | 249 | 73 | |
| XM_013071090.2 | XP_012926544.1 | 1 - 2 | 249 | 73 | ||
| XM_013071091.2 | XP_012926545.1 | 1 - 2 | 249 | 73 | ||
More Information
Set Membership: |
|

Ccnh
Genomic Map