Assay Details
| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842200.3 | XP_004842257.1 | 2 - 3 | 454 | 76 | |
| XM_021256113.1 | XP_021111772.1 | 2 - 3 | 454 | 76 | ||
More Information
Set Membership: |
|

Hspa13
Genomic Map